MilliporeSigma
  • DGKζ deficiency protects against peripheral insulin resistance and improves energy metabolism.
DGKζ deficiency protects against peripheral insulin resistance and improves energy metabolism.
Journal of lipid research (2017-10-27)
Boubacar Benziane, Melissa L Borg, Robby Z Tom, Isabelle Riedl, Julie Massart, Marie Björnholm, Marc Gilbert, Alexander V Chibalin, Juleen R Zierath
ABSTRACT
Diacylglycerol kinases (DGKs) regulate the balance between diacylglycerol (DAG) and phosphatidic acid. DGKζ is highly abundant in skeletal muscle and induces fiber hypertrophy. We hypothesized that DGKζ influences functional and metabolic adaptations in skeletal muscle and whole-body fuel utilization. DAG content was increased in skeletal muscle and adipose tissue, but unaltered in liver of DGKζ KO mice. Linear growth, body weight, fat mass, and lean mass were reduced in DGKζ KO versus wild-type mice. Conversely, male DGKζ KO and wild-type mice displayed a similar robust increase in plantaris weight after functional overload, suggesting that DGKζ is dispensable for muscle hypertrophy. Although glucose tolerance was similar, insulin levels were reduced in high-fat diet (HFD)-fed DGKζ KO versus wild-type mice. Submaximal insulin-stimulated glucose transport and p-Akt Ser473 were increased, suggesting enhanced skeletal muscle insulin sensitivity. Energy homeostasis was altered in DGKζ KO mice, as evidenced by an elevated respiratory exchange ratio, independent of altered physical activity or food intake. In conclusion, DGKζ deficiency increases tissue DAG content and leads to modest growth retardation, reduced adiposity, and protection against insulin resistance. DGKζ plays a role in the control of growth and metabolic processes, further highlighting specialized functions of DGK isoforms in type 2 diabetes pathophysiology.
Sigma-Aldrich
Anti-CGI-58 Antibody, serum, from rabbit
Sigma-Aldrich
Anti-FASN Mouse mAb (3F2-1F3), liquid, clone 3F2-1F3, Calbiochem®