MilliporeSigma
  • Hypoxia induces production of citrullinated proteins in human fibroblast-like synoviocytes through regulating HIF1α.
Hypoxia induces production of citrullinated proteins in human fibroblast-like synoviocytes through regulating HIF1α.
Scandinavian journal of immunology (2018-02-28)
R Yu, C Li, L Sun, L Jian, Z Ma, J Zhao, X Liu
ABSTRACT
Hypoxia is a prominent microenvironment feature in a range of disorders including cancer, rheumatoid arthritis (RA), atherosclerosis, inflammatory bowel disease (IBD), infection and obesity. Hypoxia promotes biological functions of fibroblast-like synoviocytes via regulating hypoxia-inducible factor 1α (HIF1α). Dysregulated protein citrullination in RA drives the production of antibodies to citrullinated proteins, a highly specific biomarker of RA. However, the mechanisms promoting citrullination in RA are not yet fully elucidated. In this study, we investigated whether pathophysiological hypoxia as found in the rheumatoid synovium modulates the citrullination in human fibroblast-like synoviocytes (HFLS). Here, we found that peptidylarginine deiminase 2 (PAD2) and citrullinated proteins were increased in HFLS after exposure to hypoxia. Moreover, knocking down HIF1α by HIF1α siRNA ameliorated the expression of PAD2 and citrullinated proteins. Collectively, this study provides a new mechanism involved in generating citrullinated proteins: hypoxia promotes citrullination and PAD production in HFLS. Concurrently, we also proposed a novel hypoxia involved mechanism in RA pathogenesis. This study deepens our understanding of the role of hypoxia in the pathogenesis of RA and provides a potential therapeutic strategy for RA.
Sigma-Aldrich
Anti-Citrulline Antibody, Upstate®, from rabbit
Pricing and availability is not currently available.