MilliporeSigma
  • Progerin, the protein responsible for the Hutchinson-Gilford progeria syndrome, increases the unrepaired DNA damages following exposure to ionizing radiation.
Progerin, the protein responsible for the Hutchinson-Gilford progeria syndrome, increases the unrepaired DNA damages following exposure to ionizing radiation.
Genes and environment : the official journal of the Japanese Environmental Mutagen Society (2015-01-01)
Asao Noda, Shuji Mishima, Yuko Hirai, Kanya Hamasaki, Reid D Landes, Hiroshi Mitani, Kei Haga, Tohru Kiyono, Nori Nakamura, Yoshiaki Kodama
ABSTRACT
Progerin, the protein responsible for the Hutchinson-Gilford Progeria Syndrome (HGPS), is a partially deleted form of nuclear lamin A, and its expression has been suggested as a cause for dysfunctional nuclear membrane and premature senescence. To examine the role of nuclear envelop architecture in regulating cellular aging and DNA repair, we used ionizing radiation to increase the number of DNA double strand breaks (DSBs) in normal and HGPS cells, and analyzed possible relationship between unrepaired DSBs and cellular aging. We found that HGPS cells are normal in repairing a major fraction of radiation-induced double strand breaks (M-DSBs)but abnormal to show increased amount of residual unrepaired DSBs (R-DSBs). Such unrepaired DSBs were 2.6 times (CI 95 %: 2.2-3.2) higher than that in normal cells one week after the irradiation, and 1.6 times (CI 95 %: 1.3-1.9) higher even one month after the irradiation. These damages tend to increase as the nuclear envelope become abnormal, a characteristic of both HGPS and normal human cells which undergo replicative senescence. The artificial, enforced over-expression of progerin further impaired the repair of M-DSBs, implying lamin A-associated nuclear membrane has an important role for DNA DSB repair. Introduction of telomerase gene function in HGPS cells reversed such aging phenotypes along with upregulation of lamin B1 and downregulation of progerin, which is a hallmark of young cells. We suggest that lamin A- or progerin-associated nuclear envelope is involved in cellular aging associated with DNA damage repair.
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-Progerin Antibody, clone 13A4, clone 13A4, Upstate®, from mouse