MilliporeSigma
  • PRDM12 Is Required for Initiation of the Nociceptive Neuron Lineage during Neurogenesis.
PRDM12 Is Required for Initiation of the Nociceptive Neuron Lineage during Neurogenesis.
Cell reports (2019-03-28)
Luca Bartesaghi, Yiqiao Wang, Paula Fontanet, Simone Wanderoy, Finja Berger, Haohao Wu, Natalia Akkuratova, Filipa Bouçanova, Jean-Jacques Médard, Charles Petitpré, Mark A Landy, Ming-Dong Zhang, Philip Harrer, Claudia Stendel, Rolf Stucka, Marina Dusl, Maria Eleni Kastriti, Laura Croci, Helen C Lai, Gian Giacomo Consalez, Alexandre Pattyn, Patrik Ernfors, Jan Senderek, Igor Adameyko, Francois Lallemend, Saida Hadjab, Roman Chrast
ABSTRACT
The sensation of pain is essential for the preservation of the functional integrity of the body. However, the key molecular regulators necessary for the initiation of the development of pain-sensing neurons have remained largely unknown. Here, we report that, in mice, inactivation of the transcriptional regulator PRDM12, which is essential for pain perception in humans, results in a complete absence of the nociceptive lineage, while proprioceptive and touch-sensitive neurons remain. Mechanistically, our data reveal that PRDM12 is required for initiation of neurogenesis and activation of a cascade of downstream pro-neuronal transcription factors, including NEUROD1, BRN3A, and ISL1, in the nociceptive lineage while it represses alternative fates other than nociceptors in progenitor cells. Our results thus demonstrate that PRDM12 is necessary for the generation of the entire lineage of pain-initiating neurons.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-PRDM12 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-Neurofilament 200 (Phos. and Non-Phos.) antibody produced in mouse, clone N52, ascites fluid
Sigma-Aldrich
Anti-Peripherin Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit