MilliporeSigma
  • Structural dynamics of the human COP9 signalosome revealed by cross-linking mass spectrometry and integrative modeling.
Structural dynamics of the human COP9 signalosome revealed by cross-linking mass spectrometry and integrative modeling.
Proceedings of the National Academy of Sciences of the United States of America (2020-02-09)
Craig Gutierrez, Ilan E Chemmama, Haibin Mao, Clinton Yu, Ignacia Echeverria, Sarah A Block, Scott D Rychnovsky, Ning Zheng, Andrej Sali, Lan Huang
ABSTRACT
The COP9 signalosome (CSN) is an evolutionarily conserved eight-subunit (CSN1-8) protein complex that controls protein ubiquitination by deneddylating Cullin-RING E3 ligases (CRLs). The activation and function of CSN hinges on its structural dynamics, which has been challenging to decipher by conventional tools. Here, we have developed a multichemistry cross-linking mass spectrometry approach enabled by three mass spectometry-cleavable cross-linkers to generate highly reliable cross-link data. We applied this approach with integrative structure modeling to determine the interaction and structural dynamics of CSN with the recently discovered ninth subunit, CSN9, in solution. Our results determined the localization of CSN9 binding sites and revealed CSN9-dependent structural changes of CSN. Together with biochemical analysis, we propose a structural model in which CSN9 binding triggers CSN to adopt a configuration that facilitates CSN-CRL interactions, thereby augmenting CSN deneddylase activity. Our integrative structure analysis workflow can be generalized to define in-solution architectures of dynamic protein complexes that remain inaccessible to other approaches.
Sigma-Aldrich
4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium tetrafluoroborate, 97%
Sigma-Aldrich
BMSO crosslinker, ≥95%
Sigma-Aldrich
DHSO crosslinker