- Exploration of a Chiral Cobalt Catalyst for Visible-Light-Induced Enantioselective Radical Conjugate Addition.
Exploration of a Chiral Cobalt Catalyst for Visible-Light-Induced Enantioselective Radical Conjugate Addition.
Angewandte Chemie (International ed. in English) (2019-07-12)
Kai Zhang, Liang-Qiu Lu, Yue Jia, Ying Wang, Fu-Dong Lu, Fangfang Pan, Wen-Jing Xiao
PMID31293048
ABSTRACT
Chiral catalysts tolerating photochemical reactions are in great demand for the vast development of visible-light-induced asymmetric synthesis. Now, chiral octahedral complexes based on earth-abundant metal and chiral N4 ligands are reported. One well-defined chiral CoII -complex is shown to be an efficient catalyst in the visible-light-induced conjugated addition of enones by alkyl and acyl radicals, providing synthetically valued chiral ketones and 1,4-dicarbonyls in 47->99 % yields with up to 97:3 e.r.