MilliporeSigma
  • Photoredox-catalyzed hydrosulfonylation reaction of electron-deficient alkenes with substituted Hantzsch esters and sulfur dioxide.
Photoredox-catalyzed hydrosulfonylation reaction of electron-deficient alkenes with substituted Hantzsch esters and sulfur dioxide.
Chemical communications (Cambridge, England) (2019-05-08)
Xuefeng Wang, Min Yang, Wenlin Xie, Xiaona Fan, Jie Wu
ABSTRACT
A sulfonylation reaction of 4-substituted Hantzsch esters, DABCO·(SO2)2, and electron-deficient alkenes at room temperature in the presence of photoredox catalysis under visible light irradiation is described. Not only (E)-chalcones but also (vinylsulfonyl)benzene and 2-vinylpyridine are all suitable substrates in the transformation. This sulfonylation reaction under mild conditions shows a broad substrate scope with a good functional group compatibility. A plausible mechanism is proposed, which shows that the 4-substituted Hantzsch esters would be radical reservoirs under photoredox catalysis. The alkyl radical generated in situ from the 4-substituted Hantzsch ester would be trapped by sulfur dioxide to produce an alkylsulfonyl radical, which would then undergo further transformation leading to the final outcome.