MilliporeSigma
  • Photoredox/Nickel-Catalyzed Single-Electron Tsuji-Trost Reaction: Development and Mechanistic Insights.
Photoredox/Nickel-Catalyzed Single-Electron Tsuji-Trost Reaction: Development and Mechanistic Insights.
Angewandte Chemie (International ed. in English) (2018-10-12)
Jennifer K Matsui, Álvaro Gutiérrez-Bonet, Madeline Rotella, Rauful Alam, Osvaldo Gutierrez, Gary A Molander
ABSTRACT
A regioselective, nickel-catalyzed photoredox allylation of secondary, benzyl, and α-alkoxy radical precursors is disclosed. Through this manifold, a variety of linear allylic alcohols and allylated monosaccharides are accessible in high yields under mild reaction conditions. Quantum mechanical calculations [DFT and DLPNO-CCSD(T)] support the mechanistic hypothesis of a Ni0 to NiII oxidative addition pathway followed by radical addition and inner-sphere allylation.