MilliporeSigma
  • Parallel Signaling through IRE1α and PERK Regulates Pancreatic Neuroendocrine Tumor Growth and Survival.
Parallel Signaling through IRE1α and PERK Regulates Pancreatic Neuroendocrine Tumor Growth and Survival.
Cancer research (2019-11-02)
Paul C Moore, Jenny Y Qi, Maike Thamsen, Rajarshi Ghosh, Justin Peng, Micah J Gliedt, Rosa Meza-Acevedo, Rachel E Warren, Annie Hiniker, Grace E Kim, Dustin J Maly, Bradley J Backes, Feroz R Papa, Scott A Oakes
ABSTRACT
Master regulators of the unfolded protein response (UPR), IRE1α and PERK, promote adaptation or apoptosis depending on the level of endoplasmic reticulum (ER) stress. Although the UPR is activated in many cancers, its effects on tumor growth remain unclear. Derived from endocrine cells, pancreatic neuroendocrine tumors (PanNET) universally hypersecrete one or more peptide hormones, likely sensitizing these cells to high ER protein-folding stress. To assess whether targeting the UPR is a viable therapeutic strategy, we analyzed human PanNET samples and found evidence of elevated ER stress and UPR activation. Genetic and pharmacologic modulation of IRE1α and PERK in cultured cells, xenograft, and spontaneous genetic (RIP-Tag2) mouse models of PanNETs revealed that UPR signaling was optimized for adaptation and that inhibiting either IRE1α or PERK led to hyperactivation and apoptotic signaling through the reciprocal arm, thereby halting tumor growth and survival. These results provide a strong rationale for therapeutically targeting the UPR in PanNETs and other cancers with elevated ER stress. SIGNIFICANCE: The UPR is upregulated in pancreatic neuroendocrine tumors and its inhibition significantly reduces tumor growth in preclinical models, providing strong rationale for targeting the UPR in these cancers.
Sigma-Aldrich
Tunicamycin from Streptomyces sp.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-c-Myc antibody, Mouse monoclonal, clone 9E10, purified from hybridoma cell culture
Sigma-Aldrich
Doxycycline hyclate
Sigma-Aldrich
Amylase Activity Assay Kit, sufficient for 100 colorimetric tests
Sigma-Aldrich
KIRA8, ≥98% (HPLC)
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film