MilliporeSigma
Mechanisms of Convergent Egg Provisioning in Poison Frogs.
Current biology : CB (2019-11-26)
Eva K Fischer, Alexandre B Roland, Nora A Moskowitz, Charles Vidoudez, Ndimbintsoa Ranaivorazo, Elicio E Tapia, Sunia A Trauger, Miguel Vences, Luis A Coloma, Lauren A O'Connell
ABSTRACT
Parental provisioning of offspring with physiological products (nursing) occurs in many animals, yet little is known about the neuroendocrine basis of nursing in non-mammalian species. Within amphibians, maternal provisioning has evolved multiple times, with mothers of some species feeding unfertilized eggs to their developing offspring until tadpoles complete metamorphosis [1-3]. We conducted field studies in Ecuador and Madagascar to ask whether convergence at the behavioral level provides similar benefits to offspring and relies on shared neural mechanisms in dendrobatid and mantellid poison frogs. At an ecological level, we found that nursing allows poison frogs to provide chemical defenses to their tadpoles in both species. At the neural level, nursing was associated with increased activity in the lateral septum and preoptic area, demonstrating recruitment of shared brain regions in the convergent evolution of nursing within frogs and across vertebrates [4]. In contrast, only mantellids showed increased oxytocin neuron activity akin to that in nursing mammals [5], suggesting evolutionary versatility in molecular mechanisms. Our findings demonstrate that maternal provisioning provides similar potential benefits to offspring and relies on similar brain regions in poison frog species with convergently evolved toxicity and maternal care. VIDEO ABSTRACT.
Sigma-Aldrich
DL-Nicotine-(methyl-d3), 99 atom % D
Sigma-Aldrich
Anti-Oxytocin Antibody, clone 4G11, ascites fluid, clone 4G11, Chemicon®