MilliporeSigma
  • Nicotinic ACh receptor α7 inhibits PDGF-induced migration of vascular smooth muscle cells by activating mitochondrial deacetylase sirtuin 3.
Nicotinic ACh receptor α7 inhibits PDGF-induced migration of vascular smooth muscle cells by activating mitochondrial deacetylase sirtuin 3.
British journal of pharmacology (2018-10-03)
Dong-Jie Li, Jie Tong, Fei-Yan Zeng, Mengqi Guo, Yong-Hua Li, Hongbo Wang, Pei Wang
ABSTRACT
PDGF-BB is an angiogenic factor involved in cardiovascular diseases. Here, we investigated the possible effects of activation of the nicotinic ACh receptor α7 subtype (α7nAChR) on PDGF-BB-induced proliferation and migration in vascular smooth muscle cells (VSMCs). PNU-282987, a selective α7nAChR pharmacological agonist, was used to activate α7nAChR. The influences of α7nAChR activation on PDGF-BB-induced proliferation and migration, as well as the phosphorylation of focal adhesion kinase (FAK)/Src, a pro-migration signalling pathway, were determined in VSMCs. A variety of biochemical assays were applied to explore the underlying molecular mechanisms. PDGF-BB induced pronounced migration and proliferation of VSMCs. Activation of α7nAChRs by PNU-282987 blocked PDGF-BB-induced migration but not proliferation in wild-type (WT) VSMCs, whereas this effect was absent in α7nAChR-knockout VSMCs. Accordingly, PNU-282987 attenuated PDGF-BB-induced phosphorylation of FAKTyr397 and SrcTyr416 in WT VSMCs. Mechanistically, PNU-282987 suppressed PDGF-BB-induced oxidative stress, as demonstrated by the alterations in ROS, H2 O2 content, superoxide anion and total antioxidant activity. A sirtuin 3 (SIRT3) inhibitor 3-(1H-1,2,3-triazol-4-yl) pyridine or shRNA-mediated SIRT3 knockdown abolished the inhibitory effect of PNU-282987. PNU-282987 did not modulate SIRT3 protein expression but enhanced mitochondrial SIRT3 deacetylase activity. In line with this action, PNU-282987 enhanced the deacetylation of mitochondrial FoxO3. Lastly, PNU-282987 corrected the PDGF-BB-induced mitochondrial dysfunction by increasing mitochondrial citrate synthase activity, ATP content and nicotinamide adenine dinucleotide pool. Pharmacological activation of α7nAChRs inhibits PDGF-BB-induced VSMC migration by activating the mitochondrial deacetylase SIRT3, implying an important role for α7nAChRs in mitochondria biology and PDGF-related diseases. This article is part of a themed section on Mitochondrial Pharmacology: Featured Mechanisms and Approaches for Therapy Translation. To view the other articles in this section visit http://onlinelibrary.wiley.com/doi/10.1111/bph.v176.22/issuetoc.
Sigma-Aldrich
Sodium 4-hydroxybenzenesulfonate dihydrate, 98%
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
Methyllycaconitine citrate salt, from Delphinium brownii seeds, ≥96% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human SIRT3