MilliporeSigma
  • Teleost IgD+IgM- B Cells Mount Clonally Expanded and Mildly Mutated Intestinal IgD Responses in the Absence of Lymphoid Follicles.
Teleost IgD+IgM- B Cells Mount Clonally Expanded and Mildly Mutated Intestinal IgD Responses in the Absence of Lymphoid Follicles.
Cell reports (2019-12-26)
Pedro Perdiguero, Alba Martín-Martín, Ottavia Benedicenti, Patricia Díaz-Rosales, Esther Morel, Estefanía Muñoz-Atienza, Mónica García-Flores, Rocío Simón, Irene Soleto, Andrea Cerutti, Carolina Tafalla
ABSTRACT
Immunoglobulin D (IgD) is an ancient antibody with dual membrane-bound and fluid-phase antigen receptor functions. The biology of secreted IgD remains elusive. Here, we demonstrate that teleost IgD+IgM- plasmablasts constitute a major lymphocyte population in some mucosal surfaces, including the gut mucosa. Remarkably, secreted IgD binds to gut commensal bacteria, which in turn stimulate IgD gene transcription in gut B cells. Accordingly, secreted IgD from gut as well as gill mucosae, but not the spleen, show a V(D)J gene configuration consistent with microbiota-driven clonal expansion and diversification, including mild somatic hypermutation. By showing that secreted IgD establishes a mutualistic relationship with commensals, our findings suggest that secreted IgD may play an evolutionary conserved role in mucosal homeostasis.
Sigma-Aldrich
Benzocaine, ≥99% (HPLC)
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Fluoromount Aqueous Mounting Medium, for use with fluorescent dye-stained tissues
Sigma-Aldrich
Oxolinic acid, quinolone antibiotic
Sigma-Aldrich
Amoxicillin, 95.0-102.0% anhydrous basis
Sigma-Aldrich
Poly-L-lysine solution, 0.01%, sterile-filtered, BioReagent, suitable for cell culture