MilliporeSigma
  • The spinster homolog, two of hearts, is required for sphingosine 1-phosphate signaling in zebrafish.
The spinster homolog, two of hearts, is required for sphingosine 1-phosphate signaling in zebrafish.
Current biology : CB (2008-12-09)
Nick Osborne, Koroboshka Brand-Arzamendi, Elke A Ober, Suk-Won Jin, Heather Verkade, Nathalia Glickman Holtzman, Deborah Yelon, Didier Y R Stainier
ABSTRACT
The bioactive lipid sphingosine 1-phosphate (S1P) and its G protein-coupled receptors play critical roles in cardiovascular, immunological, and neural development and function. Despite its importance, many questions remain about S1P signaling, including how S1P, which is synthesized intracellularly, is released from cells. Mutations in the zebrafish gene encoding the S1P receptor Miles Apart (Mil)/S1P(2) disrupt the formation of the primitive heart tube. We find that mutations of another zebrafish locus, two of hearts (toh), cause phenotypes that are morphologically indistinguishable from those seen in mil/s1p2 mutants. Positional cloning of toh reveals that it encodes a member of the Spinster-like family of putative transmembrane transporters. The biological functions of these proteins are poorly understood, although phenotypes of the Drosophila spinster and zebrafish not really started mutants suggest that these proteins may play a role in lipid trafficking. Through gain- and loss-of-function analyses, we show that toh is required for signaling by S1P(2). Further evidence indicates that Toh is involved in the trafficking or cellular release of S1P.
Sigma-Aldrich
Sphingosine 1-phosphate, ≥95%, powder
Sigma-Aldrich
Albumin, Bovine Serum, Fraction V, Fatty Acid-Free