MilliporeSigma
  • Dahuang Fuzi Decoction Attenuates Renal Fibrosis and Ameliorates Mitochondrial Dysfunction in Chronic Aristolochic Acid Nephropathy.
Dahuang Fuzi Decoction Attenuates Renal Fibrosis and Ameliorates Mitochondrial Dysfunction in Chronic Aristolochic Acid Nephropathy.
Evidence-based complementary and alternative medicine : eCAM (2017-04-20)
Guang-Xing Shui, Dong Sang, Xun Yin, Yun Cai, Wei Sun
ABSTRACT
Objectives. The effects of the traditional formula Dahuang Fuzi Decoction (DFD) on chronic aristolochic acid nephropathy (AAN) in mice and its underlying mechanisms were studied. Methods. Mice were randomly divided into the following six groups: the control group, the model group (AAN), the saline-treated group (AAN + vehicle), the normal dose DFD-treated group (AAN + NDFD), the high dose DFD-treated group (AAN + HDFD), and the rosiglitazone treated group (AAN + Rosi). After treating for 8 weeks, 24 h urine and blood samples were collected and the mice sacrificed to study the biochemical parameters associated with renal function. The samples were analyzed for renal fibrosis and mitochondrial dysfunction (MtD) markers. To achieve that, collagen III, collagen I, mitochondrial DNA copy numbers (mtDNA), mitochondrial membrane potential (MMP), ATP content, and ROS production were evaluated. Results. Our results showed that proteinuria, kidney function, and the renal pathological characteristics were improved by DFD and rosiglitazone. The expression of collagen III and collagen I decreased after treating with either DFD or rosiglitazone. Mitochondrial dysfunction based on the increase in ROS production, decrease in mitochondrial DNA copy numbers, and reduction of MMP and ATP content was improved by DFD and rosiglitazone. Conclusions. DFD could protect against renal impairments and ameliorate mitochondrial dysfunction in chronic AAN mice.
Sigma-Aldrich
Aristolochic acid I, powder