MilliporeSigma
  • PPARalpha ligands inhibit radiation-induced microglial inflammatory responses by negatively regulating NF-kappaB and AP-1 pathways.
PPARalpha ligands inhibit radiation-induced microglial inflammatory responses by negatively regulating NF-kappaB and AP-1 pathways.
Free radical biology & medicine (2008-10-15)
Sriram Ramanan, Mitra Kooshki, Weiling Zhao, Fang-Chi Hsu, Mike E Robbins
ABSTRACT
Whole-brain irradiation (WBI) can lead to cognitive impairment several months to years after irradiation. Studies on rodents have shown a rapid and sustained increase in activated microglia (brain macrophages) following brain irradiation, contributing to a chronic inflammatory response and a corresponding decrease in hippocampal neurogenesis. Thus, alleviating microglial activation following radiation represents a key strategy to minimize WBI-induced morbidity. We hypothesized that pretreatment with peroxisomal proliferator-activated receptor (PPAR)alpha agonists would ameliorate the proinflammatory responses seen in the microglia following in vitro radiation. Irradiating BV-2 cells (a murine microglial cell line) with single doses (2-10 Gy) of (137)Cs gamma-rays led to increases in (1) the gene expression of IL-1beta and TNFalpha, (2) Cox-2 protein levels, and (3) intracellular ROS generation. In addition, an increase in the DNA-binding activity of redox-regulated proinflammatory transcription factors AP-1 and NF-kappaB was observed. Pretreating BV-2 cells with the PPARalpha agonists GW7647 and Fenofibrate significantly inhibited the radiation-induced microglial proinflammatory response, in part, via decreasing (i) the nuclear translocation of the NF-kappaB p65 subunit and (ii) phosphorylation of the c-jun subunit of AP-1 in the nucleus. Taken together, these data support the hypothesis that activation of PPARalpha can modulate the radiation-induced microglial proinflammatory response.
Sigma-Aldrich
BAY 11-7082, The BAY 11-7082, also referenced under CAS 19542-67-7, controls the biological activity of TNF-α. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.
Sigma-Aldrich
NF-κB Activation Inhibitor, The NF-κB Activation Inhibitor, also referenced under CAS 545380-34-5, controls the biological activity of NF-κB. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.
Sigma-Aldrich
JNK Inhibitor II, JNK Inhibitor II. SP600125, CAS 129-56-6, is a potent, cell-permeable, selective, and ATP competitive inhibitor of c-Jun N-terminal kinase (JNK; IC₅₀ = 40 nM for JNK-1 & JNK-2 & 90 nM for JNK-3).
Sigma-Aldrich
GW7647, ≥98% (HPLC)