MilliporeSigma
  • Shogaols at proapoptotic concentrations induce G(2)/M arrest and aberrant mitotic cell death associated with tubulin aggregation.
Shogaols at proapoptotic concentrations induce G(2)/M arrest and aberrant mitotic cell death associated with tubulin aggregation.
Apoptosis : an international journal on programmed cell death (2011-05-21)
Fei-Fei Gan, Amrita A Nagle, Xiaohui Ang, Olivia Huixian Ho, Sock-Hoon Tan, Hong Yang, Wai-Keung Chui, Eng-Hui Chew
ABSTRACT
Shogaols have been previously reported to induce cancer cell death via multiple mechanisms, among which one analog 6-shogaol has been reported to cause microtubule damage through specific reaction with sulfhydryl groups in tubulin. In this study, a series of shogaols with different side chain lengths (4-, 6-, 8- and 10-shogaol) was synthesized and evaluated for antiproliferative activity in HCT 116 colon carcinoma and SH-SY5Y neuroblastoma cells. 4- and 6-shogaol were identified as lead compounds possessing the strongest antiproliferative activity. In the soft agar assay, the lead shogaols displayed dose-dependent inhibition on cancer cell colony formation under anchorage-independent conditions. Using HCT 116 as the selected cancer cell line, the molecular events linking shogaols-induced G(2)/M cell cycle arrest to apoptosis characterized by caspase 3 and PARP cleavage were investigated. At sublethal concentrations, the halt at G(2)/M phase was alleviated along time and cells survived. Conversely, proapoptotic concentrations of 4- and 6-shogaol induced irreversible G(2)/M arrest that was at least in part associated with down-regulation of cell cycle checkpoint proteins cdk1, cyclin B and cdc25C, as well as spindle assembly checkpoint proteins mad2, cdc20 and survivin. A dose- and time-dependent accumulation of insoluble tubulin in the insoluble fractions of cell lysates provided evidence that G(2) checkpoint failure led to disruption of microtubule turnover. In summary, our results conclude that shogaols cause apoptosis by inducing aberrant mitosis at least through the attenuation of cell cycle and spindle assembly checkpoint proteins.
Product Description
Supelco
[6]-Shogaol, analytical standard