MilliporeSigma
RNA SHAPE analysis in living cells.
Nature chemical biology (2012-11-28)
Robert C Spitale, Pete Crisalli, Ryan A Flynn, Eduardo A Torre, Eric T Kool, Howard Y Chang
ABSTRACT
RNA structure has important roles in practically every facet of gene regulation, but the paucity of in vivo structural probes limits current understanding. Here we design, synthesize and demonstrate two new chemical probes that enable selective 2'-hydroxyl acylation analyzed by primer extension (SHAPE) in living cells. RNA structures in human, mouse, fly, yeast and bacterial cells are read out at single-nucleotide resolution, revealing tertiary contacts and RNA-protein interactions.
Sigma-Aldrich
In Vivo SHAPE Reagent for Live Cell RNA Structure Analysis, permits the analysis of RNA structure in living cells
Sigma-Aldrich
2-Methylnicotinic acid imidazolide