MilliporeSigma
  • Sphingosine 1-phosphate rescues canine LPS-induced acute lung injury and alters systemic inflammatory cytokine production in vivo.
Sphingosine 1-phosphate rescues canine LPS-induced acute lung injury and alters systemic inflammatory cytokine production in vivo.
Translational research : the journal of laboratory and clinical medicine (2008-11-18)
William S Szczepaniak, Yingze Zhang, Sarah Hagerty, Michael T Crow, Priya Kesari, Joe G N Garcia, Augustine M K Choi, Brett A Simon, Bryan J McVerry
ABSTRACT
S1P has been demonstrated to protect against the formation of lipopolysaccharide (LPS)-induced lung edema when administered concomitantly with LPS. In the current study, we sought to determine the effectiveness of S1P to attenuate lung injury in a translationally relevant canine model of ALI when administered as rescue therapy. Secondarily, we examined whether the attenuation of LPS-induced physiologic lung injury after administration of S1P was, at least in part, caused by an alteration in local and/or systemic inflammatory cytokine expression. We examined 18, 1-year-old male beagles prospectively in which we instilled bacterial LPS (2-4 mg/kg) intratracheally followed in 1 h with intravenous S1P (85 microg/kg) or vehicle and 8 h of high-tidal-volume mechanical ventilation. S1P attenuated the formation of Q(s)/Q(t) (32%), and both the presence of protein (72%) and neutrophils (95%) in BAL fluid compared with vehicle controls. Although lung tissue inflammatory cytokine production was found to vary regionally throughout the LPS-injured lung, S1P did not alter the expression pattern. Similarly, BAL cytokine production was not altered significantly by intravenous S1P in this model. Interestingly, S1P potentiated the LPS-induced systemic production of 3 inflammatory cytokines, TNF-alpha (6-fold), KC (1.2-fold), and IL-6 (3-fold), without resulting in end-organ dysfunction. In conclusion, intravenous S1P reduces inflammatory lung injury when administered as rescue therapy in our canine model of LPS-induced ALI. This improvement is observed in the absence of changes in local pulmonary inflammatory cytokine production and an augmentation of systemic inflammation.
Sigma-Aldrich
Sphingosine 1-phosphate, ≥95%, powder
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by trichloroacetic acid extraction