MilliporeSigma
  • Diacylglycerol kinase δ and sphingomyelin synthase-related protein functionally interact via their sterile α motif domains.
Diacylglycerol kinase δ and sphingomyelin synthase-related protein functionally interact via their sterile α motif domains.
The Journal of biological chemistry (2020-01-26)
Chiaki Murakami, Fumi Hoshino, Hiromichi Sakai, Yasuhiro Hayashi, Atsushi Yamashita, Fumio Sakane
ABSTRACT
The δ isozyme of diacylglycerol kinase (DGKδ) plays critical roles in lipid signaling by converting diacylglycerol (DG) to phosphatidic acid (PA). We previously demonstrated that DGKδ preferably phosphorylates palmitic acid (16:0)- and/or palmitoleic acid (16:1)-containing DG molecular species, but not arachidonic acid (20:4)-containing DG species, which are recognized as DGK substrates derived from phosphatidylinositol turnover, in high glucose-stimulated myoblasts. However, little is known about the origin of these DG molecular species. DGKδ and two DG-generating enzymes, sphingomyelin synthase (SMS) 1 and SMS-related protein (SMSr), contain a sterile α motif domain (SAMD). In this study, we found that SMSr-SAMD, but not SMS1-SAMD, co-immunoprecipitates with DGKδ-SAMD. Full-length DGKδ co-precipitated with full-length SMSr more strongly than with SMS1. However, SAMD-deleted variants of SMSr and DGKδ interacted only weakly with full-length DGKδ and SMSr, respectively. These results strongly suggested that DGKδ interacts with SMSr through their respective SAMDs. To determine the functional outcomes of the relationship between DGKδ and SMSr, we used LC-MS/MS to investigate whether overexpression of DGKδ and/or SMSr in COS-7 cells alters the levels of PA species. We found that SMSr overexpression significantly enhances the production of 16:0- or 16:1-containing PA species such as 14:0/16:0-, 16:0/16:0-, 16:0/18:1-, and/or 16:1/18:1-PA in DGKδ-overexpressing COS-7 cells. Moreover, SMSr enhanced DGKδ activity via their SAMDs in vitro Taken together, these results strongly suggest that SMSr is a candidate DG-providing enzyme upstream of DGKδ and that the two enzymes represent a new pathway independent of phosphatidylinositol turnover.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Avanti
16:0-18:1 DG, 1-palmitoyl-2-oleoyl-sn-glycerol, chloroform
Avanti
16:0-18:1 DG, 1-palmitoyl-2-oleoyl-sn-glycerol, neat oil
Avanti
14:0 PA, Avanti Research - A Croda Brand