MilliporeSigma
  • The E2A-HLF oncogenic fusion protein acts through Lmo2 and Bcl-2 to immortalize hematopoietic progenitors.
The E2A-HLF oncogenic fusion protein acts through Lmo2 and Bcl-2 to immortalize hematopoietic progenitors.
Leukemia (2010-11-13)
J de Boer, J Yeung, J Ellu, R Ramanujachar, B Bornhauser, O Solarska, M Hubank, O Williams, H J M Brady
ABSTRACT
The oncogenic fusion protein E2A-HLF is a chimeric transcription factor that arises from the t(17;19) translocation in childhood B-cell acute lymphoblastic leukemias (B-precursor ALL) and is associated with very poor outcome. We show that retroviral-mediated expression of E2A-HLF alone is sufficient to immortalize primary lymphoid progenitors. We identify Lmo2 and Bcl-2 as direct target genes downstream of E2A-HLF. We use real-time PCR analysis to show that LMO2 and BCL-2 expression is preferentially upregulated both in biopsy material from t(17;19) B-precursor ALL patients and lymphoid cell lines derived from t(17;19) leukemias. Co-expression of Lmo2 and Bcl-2 was sufficient to immortalize lymphoid progenitor cells resulting in a similar phenotype to that induced by E2A-HLF alone. Both shRNA-mediated knockdown of Lmo2 expression and pharmacological inhibition of BCL-2 function in E2A-HLF immortalized cells severely compromised their viability. These data suggest that both Lmo2 and Bcl-2 are required for the action of E2A-HLF in leukemogenesis.
Sigma-Aldrich
Anti-Myc Tag Antibody, clone 4A6, clone 4A6, Upstate®, from mouse
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Integrin α2 Antibody, clone P1E6, clone P1E6, Chemicon®, from mouse
Pricing and availability is not currently available.