MilliporeSigma
  • Exploiting the transcriptional specificity of the alpha-methylacyl-CoA racemase AMACR promoter for the molecular imaging of prostate cancer.
Exploiting the transcriptional specificity of the alpha-methylacyl-CoA racemase AMACR promoter for the molecular imaging of prostate cancer.
Oncotarget (2019-01-08)
Mariya Shapovalova, Julia Davydova, Christine Henzler, Mark Daniel, Scott M Dehm, Christopher A Warlick, Aaron M LeBeau
ABSTRACT
The metabolic protein alpha-methylacyl-CoA racemase (AMACR) is significantly overexpressed in prostate cancer compared to the normal prostate and other non-malignant tissue. Though an attractive target, there are no reports in the literature on leveraging the expression of AMACR for the molecular imaging of prostate cancer. Here, we used a molecular-genetic imaging strategy to exploit the transcriptional specificity of the AMACR promoter for the in vivo detection of prostate cancer using the reporter gene luciferase. We performed a stepwise truncation of the promoter and identified a 565 base pair minimal promoter for AMACR that retained both high activity and specificity. Following identification of the minimal promoter for AMACR, we used an advanced two-step transcriptional amplification system to maximize the promoter output. We showed that our optimized AMACR promoter can drive expression of luciferase for molecular imaging in subcutaneous xenograft models of androgen receptor-positive and androgen receptor-negative prostate cancer using a non-replicative adenovirus for gene delivery. Our results provide evidence that the AMACR promoter can be exploited to drive the cancer-specific expression of reporter genes and potentially even be incorporated into conditionally replicative adenoviruses for oncolytic therapy and other applications.
Sigma-Aldrich
Anti-AMACR antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
GeneJuice® Transfection Reagent, Non-lipid based chemical transfection reagent optimized for maximum transfection efficiency, ease-of-use, and minimal cytotoxicity on a wide variety of mammalian cells.