MilliporeSigma
  • Biochemical, Biophysical and Cellular Techniques to Study the Guanine Nucleotide Exchange Factor, GIV/Girdin.
Biochemical, Biophysical and Cellular Techniques to Study the Guanine Nucleotide Exchange Factor, GIV/Girdin.
Current protocols in chemical biology (2016-12-08)
Pradipta Ghosh, Nicolas Aznar, Lee Swanson, I-Chung Lo, Inmaculada Lopez-Sanchez, Jason Ear, Cristina Rohena, Nicholas Kalogriopoulos, Linda Joosen, Ying Dunkel, Nina Sun, Peter Nguyen, Deepali Bhandari
ABSTRACT
Canonical signal transduction via heterotrimeric G proteins is spatiotemporally restricted, i.e., triggered exclusively at the plasma membrane, only by agonist activation of G protein-coupled receptors via a finite process that is terminated within a few hundred milliseconds. Recently, a rapidly emerging paradigm has revealed a noncanonical pathway for activation of heterotrimeric G proteins via the nonreceptor guanidine-nucleotide exchange factor, GIV/Girdin. Biochemical, biophysical, and functional studies evaluating this pathway have unraveled its unique properties and distinctive spatiotemporal features. As in the case of any new pathway/paradigm, these studies first required an in-depth optimization of tools/techniques and protocols, governed by rationale and fundamentals unique to the pathway, and more specifically to the large multimodular GIV protein. Here we provide the most up-to-date overview of protocols that have generated most of what we know today about noncanonical G protein activation by GIV and its relevance in health and disease. © 2016 by John Wiley & Sons, Inc.
Sigma-Aldrich
GeneJuice® Transfection Reagent, Non-lipid based chemical transfection reagent optimized for maximum transfection efficiency, ease-of-use, and minimal cytotoxicity on a wide variety of mammalian cells.
Sigma-Aldrich
Anti-Girdin Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Girdin, coiled-coil region Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Girdin Antibody, CT, clone 10E6.1, clone 10E6.1, from mouse