MilliporeSigma
  • Interaction of Vitamin K1 and Vitamin K2 with Dimyristoylphosphatidylcholine and Their Location in the Membrane.
Interaction of Vitamin K1 and Vitamin K2 with Dimyristoylphosphatidylcholine and Their Location in the Membrane.
Langmuir : the ACS journal of surfaces and colloids (2020-01-14)
Alessio Ausili, Javier Clemente, Óscar D Pons-Belda, Ana de Godos, Senena Corbalán-García, Alejandro Torrecillas, José A Teruel, Juan C Ghmez-Fernández
ABSTRACT
Vitamin K1 and vitamin K2 play very important biological roles as members of chains of electron transport as antioxidants in membranes and as cofactors for the posttranslational modification of proteins that participate in a number of physiological functions such as coagulation. The interaction of these vitamins with dimyristoylphosphatidylcholine (DMPC) model membranes has been studied by using a biophysical approach. It was observed by using differential scanning calorimetry that both vitamins have a very limited miscibility with DMPC and they form domains rich in the vitamins at high concentrations. Experiments using X-ray diffraction also showed the formation of different phases as a consequence of the inclusion of either vitamin K at temperatures below the phase transition. However, in the fluid state, a homogeneous phase was detected, and a decrease in the thickness of the membrane was accompanied by an increase in the water layer thickness. 2H NMR spectroscopy showed that both vitamins K induced a decrease in the onset of the phase transition, which was bigger for vitamin K1, and both vitamins decreased the order of the membrane as seen through the first moment (M1). 1H NOESY MAS-NMR showed that protons located at the rings or at the beginning of the lateral chain of both vitamins K interacted with a clear preference with protons located in the polar part of DMPC. On the other hand, protons located on the lateral chain have a nearer proximity with the methyl end of the myristoyl chains of DMPC. In agreement with the 2H NMR, ATR-FTIR (attenuated total reflectance Fourier transform infrared spectroscopy) indicated that both vitamins decreased the order parameters of DMPC. It was additionally deduced that the lateral chains of both vitamins were oriented almost in parallel to the myristoyl chains of the phospholipid.
Product Description
Avanti
14:0 PC-d54, 1,2-dimyristoyl-d54-sn-glycero-3-phosphocholine, powder
Avanti
14:0 PC-d54, 1,2-dimyristoyl-d54-sn-glycero-3-phosphocholine, chloroform