MilliporeSigma
  • The oncology drug elesclomol selectively transports copper to the mitochondria to induce oxidative stress in cancer cells.
The oncology drug elesclomol selectively transports copper to the mitochondria to induce oxidative stress in cancer cells.
Free radical biology & medicine (2012-05-01)
Masazumi Nagai, Nha H Vo, Luisa Shin Ogawa, Dinesh Chimmanamada, Takayo Inoue, John Chu, Britte C Beaudette-Zlatanova, Rongzhen Lu, Ronald K Blackman, James Barsoum, Keizo Koya, Yumiko Wada
ABSTRACT
Elesclomol is an investigational drug that exerts potent anticancer activity through the elevation of reactive oxygen species (ROS) levels and is currently under clinical evaluation as a novel anticancer therapeutic. Here we report the first description of selective mitochondrial ROS induction by elesclomol in cancer cells based on the unique physicochemical properties of the compound. Elesclomol preferentially chelates copper (Cu) outside of cells and enters as elesclomol-Cu(II). The elesclomol-Cu(II) complex then rapidly and selectively transports the copper to mitochondria. In this organelle Cu(II) is reduced to Cu(I), followed by subsequent ROS generation. Upon dissociation from the complex, elesclomol is effluxed from cells and repeats shuttling elesclomol-Cu complexes from the extracellular to the intracellular compartments, leading to continued copper accumulation within mitochondria. An optimal range of redox potentials exhibited by copper chelates of elesclomol and its analogs correlated with the elevation of mitochondrial Cu(I) levels and cytotoxic activity, suggesting that redox reduction of the copper triggers mitochondrial ROS induction. Importantly the mitochondrial selectivity exhibited by elesclomol is a distinct characteristic of the compound that is not shared by other chelators, including disulfiram. Together these findings highlight a unique mechanism of action with important implications for cancer therapy.
Sigma-Aldrich
Elesclomol, ≥98% (HPLC)