MilliporeSigma
  • Lack of glutathione peroxidase-8 in the ER impacts on lipid composition of HeLa cells microsomal membranes.
Lack of glutathione peroxidase-8 in the ER impacts on lipid composition of HeLa cells microsomal membranes.
Free radical biology & medicine (2019-12-21)
Valentina Bosello Travain, Giovanni Miotto, Ana-Marija Vučković, Giorgio Cozza, Antonella Roveri, Stefano Toppo, Fulvio Ursini, Rina Venerando, Mattia Zaccarin, Matilde Maiorino
ABSTRACT
GPx8 is a glutathione peroxidase homolog inserted in the membranes of endoplasmic reticulum (ER), where it seemingly plays a role in controlling redox status by preventing the spill of H2O2. We addressed the impact of GPx8 silencing on the lipidome of microsomal membranes, using stably GPx8-silenced HeLa cells. The two cell lines were clearly separated by Principal Component Analysis (PCA) and Partial Least Square Discriminant analysis (PLS-DA) of lipidome. Considering in detail the individual lipid classes, we observed that unsaturated glycerophospholipids (GPL) decreased, while only in phosphatidylinositols (PI) a substitution of monounsaturated fatty acids (MUFA) for polyunsaturated fatty acids (PUFA) was observed. Among sphingolipids (SL), ceramides (CER) decreased while sphingomyelins (SM) and neutral glycophingolipids (nGSL) increased. Here, in addition, longer chains than in controls in the amide fatty acid were present. The increase up to four folds of the CER (d18:1; c24:0) containing three hexose units, was the most remarkable species increasing in the differential lipidome of siGPx8 cells. Quantitative RT-PCR complied with lipidomic analysis specifically showing an increased expression of: i) acyl-CoA synthetase 5 (ACSL5); ii) CER synthase 2 and 4; iii) CER transporter (CERT); iv) UDP-glucosyl transferase (UDP-GlcT), associated to a decreased expression of UDP-galactosyl transferase (UDP-GalT). A role of the unfolded protein response (UPR) and the spliced form of the transcription factor XBP1 on the transcriptional changes of GPx8 silenced cells was ruled-out. Similarly, also the involvement of Nrf2 and NF-κB. Altogether our results indicate that GPx8-silencing of HeLa yields a membrane depleted by about 24% of polyunsaturated GPL and a corresponding increase of saturated or monounsaturated SM and specific nGSL. This is tentatively interpreted as an adaptive mechanism leading to an increased resistance to radical oxidations. Moreover, the marked shift of fatty acid composition of PI emerges as a possibly relevant issue in respect to the impact of GPx8 on signaling pathways.
Product Description
Avanti
Mouse SPLASH LIPIDOMIX Mass Spec Standard, Avanti Research - A Croda Brand
Avanti
Deuterated Ceramide LIPIDOMIX Mass Spec Standard, Avanti Research - A Croda Brand
Avanti
15:0 PE, 1,2-dipentadecanoyl-sn-glycero-3-phosphoethanolamine, powder