MilliporeSigma
  • Phosphatidic acid is required for the constitutive ruffling and macropinocytosis of phagocytes.
Phosphatidic acid is required for the constitutive ruffling and macropinocytosis of phagocytes.
Molecular biology of the cell (2013-04-12)
Michal Bohdanowicz, Daniel Schlam, Martin Hermansson, David Rizzuti, Gregory D Fairn, Takehiko Ueyama, Pentti Somerharju, Guangwei Du, Sergio Grinstein
ABSTRACT
Macrophages and dendritic cells continuously survey their environment in search of foreign particles and soluble antigens. Such surveillance involves the ongoing extension of actin-rich protrusions and the consequent formation of phagosomes and macropinosomes. The signals inducing this constitutive cytoskeletal remodeling have not been defined. We report that, unlike nonphagocytic cells, macrophages and immature dendritic cells have elevated levels of phosphatidic acid (PA) in their plasma membrane. The plasmalemmal PA is synthesized by phosphorylation of diacylglycerol, which is in turn generated by a G protein-stimulated phospholipase C. Inhibition of diacylglycerol kinase activity results in the detachment of T-cell lymphoma invasion and metastasis-inducing protein 1 (TIAM1)-a Rac guanine exchange factor-from the plasma membrane, thereby depressing Rac activity and abolishing the constitutive ruffling and macropinocytosis that characterize macrophages and immature dendritic cells. Accumulation of PA and binding of TIAM1 to the membrane require the activity of phosphatidylinositol-4,5-bisphosphate 3-kinase. Thus a distinctive, constitutive pathway of PA biosynthesis promotes the actin remodeling required for immune surveillance.
Product Description
Avanti
Soy PA, Avanti Research - A Croda Brand 840074P, powder
Avanti
Soy PA, Avanti Research - A Croda Brand 840074C
Avanti
08:0 DG, 1,2-dioctanoyl-sn-glycerol, chloroform solution
Avanti
08:0 DG, 1,2-dioctanoyl-sn-glycerol, neat oil