MilliporeSigma
  • Phospholipase D signaling in serotonin-induced mitogenesis of pulmonary artery smooth muscle cells.
Phospholipase D signaling in serotonin-induced mitogenesis of pulmonary artery smooth muscle cells.
American journal of physiology. Lung cellular and molecular physiology (2008-07-16)
Y Liu, B L Fanburg
ABSTRACT
We have previously reported the participation of mitogen-activated protein, Rho, and phosphoinositide-3 (PI3) kinases in separate pathways in serotonin (5-HT)-induced proliferation of pulmonary artery smooth muscle cells (SMCs). In this study, we investigated the possible participation of phospholipase D (PLD) and phosphatidic acid (PA) in this growth process. 5-HT stimulated a time-dependent increase in [(3)H]phosphatidylbutanol and PA generation. Exposure of SMCs to 1-butanol or overexpression of an inactive mutant of human PLD1R898R blocked 5-HT-induced proliferation. Furthermore, 1-butanol inhibited 5-HT activation of S6K1 and S6 protein, downstream effectors of mammalian target of rapamycin (mTOR), by 80 and 72%, respectively, and partially blocked activation of extracellular signal-regulated kinase (ERK) by 30% but had no effect on other associated signaling pathways. Exogenous PA caused cellular proliferation and revitalized cyclin D1 expression by 5-HT of the 1-butanol-treated cells. PA also reproduced activations by 5-HT of mTOR, S6K1, and ERK. Transfection with inactive human PLD1 reduced 5-HT-induced activation of S6K1 by approximately 50%. Inhibition of 5-HT receptor 2A (R 2A) with ketaserin blocked PLD activation by 5-HT. Inhibition with PI3-kinase inhibitor failed to block either activation of PLD by 5-HT or PA-dependent S6K1 phosphorylation. Taken together, these results indicate that ligation of the 5-HTR 2A by 5-HT initiates PLD activation in SMCs, and that its product, PA, is an early signaling molecule in 5-HT-induced pulmonary artery SMC proliferation. Signaling by PA produces its downstream effects primarily through the mTOR/S6K1 pathway and to a lesser extent through the ERK pathway. Hydrolysis of cell membrane lipid may be important in vascular effects of 5-HT.
Product Description
Avanti
18:1 Phosphatidylbutanol, 1,2-dioleoyl-sn-glycero-3-phosphobutanol (sodium salt), neat oil
Avanti
18:0-18:1 PA, 1-stearoyl-2-oleoyl-sn-glycero-3-phosphate (sodium salt), chloroform
Avanti
18:0-18:1 PA, 1-stearoyl-2-oleoyl-sn-glycero-3-phosphate (sodium salt), powder