MilliporeSigma
  • Highly enantioselective nickel-catalyzed intramolecular reductive cyclization of alkynones.
Highly enantioselective nickel-catalyzed intramolecular reductive cyclization of alkynones.
Angewandte Chemie (International ed. in English) (2015-01-20)
Wenzhen Fu, Ming Nie, Aizhen Wang, Ziping Cao, Wenjun Tang
ABSTRACT
The first asymmetric nickel-catalyzed intramolecular reductive cyclization of alkynones is reported. A P-chiral monophosphine and triethylsilane were used as the ligand and the reducing reagent, respectively, to form a series of tertiary allylic alcohols bearing furan/pyran rings in excellent yields and enantioselectivities. This reaction has a broad substrate scope and enabled the efficient synthesis of dehydroxycubebin and chiral dibenzocyclooctadiene skeletons.
Sigma-Aldrich
(S)-BIDIME, ≥97%
Sigma-Aldrich
(S)-AntPhos, ≥97%
Sigma-Aldrich
(R)-BIDIME, ≥97%