MilliporeSigma
  • Dectin-1 Binding to Annexins on Apoptotic Cells Induces Peripheral Immune Tolerance via NADPH Oxidase-2.
Dectin-1 Binding to Annexins on Apoptotic Cells Induces Peripheral Immune Tolerance via NADPH Oxidase-2.
Cell reports (2019-12-26)
Kevin Bode, Fatmire Bujupi, Corinna Link, Tobias Hein, Stephanie Zimmermann, Diluka Peiris, Vincent Jaquet, Bernd Lepenies, Heiko Weyd, Peter H Krammer
ABSTRACT
Uptake of apoptotic cells (ACs) by dendritic cells (DCs) and induction of a tolerogenic DC phenotype is an important mechanism for establishing peripheral tolerance to self-antigens. The receptors involved and underlying signaling pathways are not fully understood. Here, we identify Dectin-1 as a crucial tolerogenic receptor binding with nanomolar affinity to the core domain of several annexins (annexin A1, A5, and A13) exposed on ACs. Annexins bind to Dectin-1 on a site distinct from the interaction site of pathogen-derived β-glucans. Subsequent tolerogenic signaling induces selective phosphorylation of spleen tyrosine kinase (SYK), causing activation of NADPH oxidase-2 and moderate production of reactive oxygen species. Thus, mice deficient for Dectin-1 develop autoimmune pathologies (autoantibodies and splenomegaly) and generate stronger immune responses (cytotoxic T cells) against ACs. Our data describe an important immunological checkpoint system and provide a link between immunosuppressive signals of ACs and maintenance of peripheral immune tolerance.
Sigma-Aldrich
SYBR® Green I nucleic acid gel stain, 10,000 × in DMSO
Sigma-Aldrich
Polymyxin B sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, With MgCl2 and CaCl2, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
OPI Media Supplement, Hybri-Max, γ-irradiated, powder, suitable for hybridoma
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
2′,7′-Dichlorodihydrofluorescein diacetate, ≥97%
Avanti
18:1 Cy5 PE, Avanti Research - A Croda Brand
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Roche
DNase I, grade II, from bovine pancreas
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand