MilliporeSigma
  • High-throughput assays to measure intracellular Ca²⁺ mobilization in cells that express recombinant S1P receptor subtypes.
High-throughput assays to measure intracellular Ca²⁺ mobilization in cells that express recombinant S1P receptor subtypes.
Methods in molecular biology (Clifton, N.J.) (2012-04-25)
William J Valentine, Gabor Tigyi
ABSTRACT
Intracellular Ca(2+) mobilization is a useful readout to screen for agonists or antagonists of G-protein -coupled receptors (GPCRs). Here, we describe methods to conduct high-throughput screening of stably or transiently transfected HTC4 cells expressing the individual S1P1-5 receptor subtypes. The cells are grown in 96-well plates and loaded with the cell permeable fluorescent Ca(2+) indicator dye Fura-2-AM. Changes in intracellular Ca(2+) levels in response to S1P or test compounds are detected using a FlexStation II scanning fluorometer with integrated fluidics transfer capabilities.
Product Description
Avanti
3-deoxy sphingosine, Avanti Research - A Croda Brand 860469P, powder
Avanti
Sphingosine-1-Phosphate (d18:1), Avanti Research - A Croda Brand