MilliporeSigma
  • Enantioselective Synthesis of Chiral-at-Cage o-Carboranes via Pd-Catalyzed Asymmetric B-H Substitution.
Enantioselective Synthesis of Chiral-at-Cage o-Carboranes via Pd-Catalyzed Asymmetric B-H Substitution.
Journal of the American Chemical Society (2018-03-27)
Ruofei Cheng, Bowen Li, Jie Wu, Jie Zhang, Zaozao Qiu, Wenjun Tang, Shu-Li You, Yong Tang, Zuowei Xie
ABSTRACT
Carborane cage chirality is an outstanding issue of great interest as the icosahedral carboranes have wide applications in medicinal and materials chemistry. The synthesis of optically active carborane derivatives, whose chirality is associated with the substitution patterns on the polyhedron, will open new avenues to carborane chemistry. We report herein an efficient method to achieve chiral-at-cage arylation of o-carboranes with high regio- and enantioselectivities by a strategy of palladium-catalyzed asymmetric intramolecular B-H arylation and cyclization. This represents the first example of the enantioselective reaction on carboranes, providing an efficient way for the construction of chiral-at-cage compounds with new skeletons.
Sigma-Aldrich
(R)-BIDIME, ≥97%
Sigma-Aldrich
(R)-AntPhos, ≥97%
Sigma-Aldrich
(S)-AntPhos, ≥97%
Sigma-Aldrich
(S)-BIDIME, ≥97%