MilliporeSigma
  • Reduced Toxicity of Liposomal Nitrogen Mustard Prodrug Formulation Activated by an Intracellular ROS Feedback Mechanism in Hematological Neoplasm Models.
Reduced Toxicity of Liposomal Nitrogen Mustard Prodrug Formulation Activated by an Intracellular ROS Feedback Mechanism in Hematological Neoplasm Models.
Molecular pharmaceutics (2019-12-12)
Mengting Lin, Wangwei Guo, Zhentao Zhang, Yi Zhou, Jiejian Chen, Tiantian Wang, Xincheng Zhong, Yiying Lu, Qiyao Yang, Qichun Wei, Min Han, Donghang Xu, Jianqing Gao
ABSTRACT
Nitrogen mustard (NM) is among the earliest drugs used to treat malignant tumors and it kills tumor cells by cross-linking DNA. Unfortunately, because of the short half-life and unfavorable selectivity, NM causes significant damage to normal tissues. As NM can increase the levels of reactive oxygen species (ROS) in tumor cells, a ROS-activated nitrogen mustard prodrug (NM-Pro) was synthesized and mixed with NM at a specific ratio to obtain an "NM-ROS-NM-Pro-NM" positive feedback system, which ultimately achieves a specific lethal effect on hematological neoplasms. The further encapsulation of NM/NM-Pro in liposomes allows the sustained release of the drug and prolongs the residence time in vivo. Here, we prepared stable liposomes with a uniform particle size of 170.6 ± 2.2 nm. The optimal ratio of NM to NM-Pro in this study was 2:1. The active drug NM in the NM/NM-Pro system continuously stimulated ROS production by the cells, which in turn further activated the NM-Pro to continuously generate NM. The positive feedback pathway between the NM and NM-Pro resulted in the specific death of tumor cells. Furthermore, the K562 hematological neoplasm model was utilized to evaluate the therapeutic effect of NM/NM-Pro liposomes in vivo. After encapsulation in liposomes, the targeting of tumor cells was increased approximately two times compared with that of normal cells, and NM/NM-Pro liposomes exhibited reduced toxicity, without an increase in drug activity compared to the NM/NM-Pro combination. The NM/NM-Pro delivery system exerts a positive feedback effect on ROS production in tumor cells and displays good potential for the specific killing of hematoma cells.
Product Description
Avanti
Hydro Soy PC, Avanti Research - A Croda Brand