MilliporeSigma
  • Enzyme Promiscuity as a Remedy for the Common Problems with Knoevenagel Condensation.
Enzyme Promiscuity as a Remedy for the Common Problems with Knoevenagel Condensation.
Chemistry (Weinheim an der Bergstrasse, Germany) (2019-05-29)
Dominik Koszelewski, Ryszard Ostaszewski
ABSTRACT
A new protocol based on lipase-catalyzed tandem reaction toward α,β-enones/enoesters is presented. For the synthesis of the desired products the tandem process based on enzyme-catalyzed hydrolysis and Knoevenagel reaction starting from enol acetates and aldehyde is developed. The relevant impact of the reaction conditions including organic solvent, enzyme type, and temperature on the course of the reaction was revealed. It was shown that controllable release of the active methylene compound from the corresponding enol carboxylate ensured by enzymatic reaction diminishes significantly the formation of the unwanted co-products. Furthermore, this protocol was extended by including a second tandem chemoenzymatic transformation engaging various aldehyde precursors. After a careful optimization of the reaction conditions, the target products were obtained with yields up to 86 % and with excellent E/Z-selectivity.
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Amano Lipase from Pseudomonas fluorescens, beige-brown, ≥20,000 U/g
Sigma-Aldrich
Amano Lipase PS, from Burkholderia cepacia, ≥23,000  U/g, pH 7.0, 50 °C (Optimum pH and temperature)