MilliporeSigma
  • Tumor Cells Modified with Newcastle Disease Virus Expressing IL-24 as a Cancer Vaccine.
Tumor Cells Modified with Newcastle Disease Virus Expressing IL-24 as a Cancer Vaccine.
Molecular therapy oncolytics (2019-07-25)
Xiaojing Xu, Cheng Yi, Xiaoqin Yang, Jianwei Xu, Qing Sun, Yonghao Liu, Lixiang Zhao
ABSTRACT
Interleukin-24 (IL-24) is a promising agent for cancer immunotherapy that induces apoptosis of tumor cells and enhances T cell activation and function. In order to improve the antitumor activity induced by Newcastle disease virus (NDV)-modified tumor vaccine, we generated a recombinant NDV expressing IL-24 using reverse genetics. Irradiated tumor cells infected with LX/IL-24 showed stable IL-24 expression. The cytotoxicity assay showed that LX/IL-24-infected murine melanoma cells significantly enhanced the antitumor immune response in vitro. Then, the antitumor effects of virus-infected tumor cells were examined in the murine tumor models. LX/IL-24-infected tumor cells exhibited strong antitumor effects both in prophylaxis and therapeutic models. LX/IL-24-infected tumor cells increased infiltration of CD4+ T cells and CD8+ T cells in tumor sites, and the antitumor activity of the tumor vaccine modified with LX/IL-24 was dependent on CD8+ T cells. Taken together, our data well illustrates that LX/IL-24-modified tumor cells are a promising agent for cancer immunotherapy.
Sigma-Aldrich
Human IL24 / Interleukin-24 ELISA Kit