MilliporeSigma
  • Conjugate prodrug AN-233 induces fetal hemoglobin expression in sickle erythroid progenitors and β-YAC transgenic mice.
Conjugate prodrug AN-233 induces fetal hemoglobin expression in sickle erythroid progenitors and β-YAC transgenic mice.
Blood cells, molecules & diseases (2019-07-28)
Aluya R Oseghale, Xingguo Zhu, Biaoru Li, Kenneth R Peterson, Abraham Nudelman, Ada Rephaeli, Hongyan Xu, Betty S Pace
ABSTRACT
Pharmacologic induction of fetal hemoglobin (HbF) is an effective strategy for treating sickle cell disease (SCD) by ameliorating disease severity. Hydroxyurea is the only FDA-approved agent that induces HbF, but significant non-responders and requirement for frequent monitoring of blood counts for drug toxicity limit clinical usefulness. Therefore, we investigated a novel prodrug conjugate of butyric acid (BA) and δ-aminolevulinate (ALA) as a potential HbF inducing agent, using erythroid precursors and a preclinical β-YAC mouse model. We observed significantly increased γ-globin gene transcription and HbF expression mediated by AN-233 in K562 cells. Moreover, AN-233 stimulated mild heme biosynthesis and inhibited expression of heme-regulated eIF2α kinase involved in silencing γ-globin expression. Studies using primary erythroid precursors generated from sickle peripheral blood mononuclear cells verified the ability of AN-233 to induce HbF, increase histone H3 and H4 acetylation levels at the γ-globin promoter and reduce erythroid precursor sickling by 50%. Subsequent drug treatment of β-YAC transgenic mice confirmed HbF induction in vivo by AN-233 through an increase in the percentage of HbF positive red blood cells and HbF levels measured by flow cytometry. These data support the potential development of AN-233 for the treatment of SCD.
Sigma-Aldrich
IgG from rabbit serum, technical grade, ≥80% (SDS-PAGE), buffered aqueous solution
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)