MilliporeSigma
  • Analysis of decolorization potential of Myrothecium roridum in the light of its secretome and toxicological studies.
Analysis of decolorization potential of Myrothecium roridum in the light of its secretome and toxicological studies.
Environmental science and pollution research international (2019-07-10)
Anna Jasińska, Adrian Soboń, Aleksandra Góralczyk-Bińkowska, Jerzy Długoński
ABSTRACT
To identify the enzymes potentially useful for the decolorization of azo dyes, the secretome of the ascomycetous fungus Myrothecium roridum IM6482 was studied by using a bottom-up proteomic approach. Among the identified proteins, the most promising for dye removal was laccase, which decolorized respectively, 66, 91, 79, and 80% of Acid Blue 113 (AB 113), Acid Red 27 (AR 27), Direct Blue 14 (DB 14), and Acid Orange 7 (AO 7). The degradation of dyes was enhanced at the wide range of pH from 4 to 8. The addition of redox mediators allowed eliminating AB 113 in concentrations up to 400 mg/L and decolorization of the simulated textile effluent. Microbial toxicity and phytotoxicity tests indicated that dyes are converted into low-toxicity metabolites. This is the first insight into the M. roridum secretome, its identification and its application for removal of select azo dyes. Obtained results extended knowledge concerning biodegradative potential of ascomycetous, ligninolytic fungi and will contribute to the improvement of dye removal by fungi.
Sigma-Aldrich
Dess-Martin periodinane, 97%
Sigma-Aldrich
2,2,6,6-Tetramethylpiperidine, ≥99%