MilliporeSigma
  • DYRK1A aggravates β cell dysfunction and apoptosis by promoting the phosphorylation and degradation of IRS2.
DYRK1A aggravates β cell dysfunction and apoptosis by promoting the phosphorylation and degradation of IRS2.
Experimental gerontology (2019-07-16)
Mei Lu, Lin Ma, Peiyan Shan, Aifen Liu, Xiaolin Yu, Wenjing Jiang, Xinbang Wang, Xinjing Zhao, Xiang Ye, Tan Wang
ABSTRACT
In this study, we aimed to investigate the role of dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 1A (DYRK1A), which is one of the most important regulators of Alzheimer's disease development, in islet β cell dysfunction and apoptosis. We found significantly increased expression of DYRK1A in both the hippocampus and pancreatic islets of APPswe/PS1ΔE9 transgenic mice than in wild-type littermates. Furthermore, we observed that the overexpression of DYRK1A greatly aggravated β cell apoptosis. Most importantly, we found that DYRK1A directly interacted with insulin receptor substrate-2 (IRS2) and promoted IRS2 phosphorylation, leading to the proteasomal degradation of IRS2 and promotion of β cell dysfunction and apoptosis. These findings suggested that DYRK1A is a potential drug target in diabetes mellitus.
Sigma-Aldrich
O-Phospho-L-tyrosine
Sigma-Aldrich
Harmine, 98%
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
MISSION® esiRNA, targeting human DYRK1A