MilliporeSigma
  • Effect of IBDV infection on the interfollicular epithelium of chicken bursa of Fabricius.
Effect of IBDV infection on the interfollicular epithelium of chicken bursa of Fabricius.
Poultry science (2018-11-28)
Ildikó Bódi, Balázs Felföldi, Krisztina Minkó, Zsófia Benyeda, Nándor Nagy, Anna L Kiss, Vilmos Palya, Imre Oláh
ABSTRACT
In the chicken bursa of Fabricius (BF), the interfollicular epithelium (IFE) consists of cylindrical- and cuboidal-shaped cells. Among the cylindrical-shaped epithelial cells, mucus-producing and caveolin-1 (Cav-1)-expressing cells can be distinguished. Occasionally, the cuboidal-shaped cells also express Cav-1, which suggests that they are precursors of both mucus-producing and Cav-1-expressing cells. Very virulent infectious bursal disease virus (IBDV) impedes the differentiation of Cav-1-expressing cells and shifts the differentiation of cuboidal cells towards mucus-producing cells. In control birds exclusively, the IFE surface shows a mucous membrane, but after IBDV infection, the surfaces of both IFE and FAE are also covered by a mucous membrane. After IBDV infection, the cells of FAE also produce mucus, providing evidence for cell transformation. In late postinfection (pi; 28 d pi), the Cav-1 expression returned in the IFE cells, whereas the follicle (the primary lymphoid organ) underwent atrophy. The appearance of the renewed Cav-1-positive cells is similar to that of the normal basal cell, but they randomly locate in different levels of IFE, suggesting the loss of epithelial polarity. Between days 2 and 7 pi, the Cav-1 expression in the endothelial cells of the cortico-medullary capillary web is variable, which may explain the hemorrhage in several infected birds. The IBDV infection stops the Cav-1 expression and subsequently the cholesterol efflux into the bursal lumen. In the infected birds, the high cholesterol level may further worsen the clinical syndrome of IBDV.
Sigma-Aldrich
3,3′-Diaminobenzidine, 97% (HPLC)
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
4-Chloro-1-naphthol, crystalline