MilliporeSigma
  • Microwave assisted high performance liquid chromatography for the separation of triacylglycerols in vegetable oils using an evaporative light scattering detector.
Microwave assisted high performance liquid chromatography for the separation of triacylglycerols in vegetable oils using an evaporative light scattering detector.
Food chemistry (2019-07-23)
Silvia Carballo-Marrero, Maria Soledad Prats-Moya, Salvador E Maestre-Pérez, José Luis Todolí-Torro
ABSTRACT
Microwave (MW) radiation was applied to perform the separation of triacylglycerols (TGs) in oil samples. The novelty of the work lies in the application of MW radiation to assist the separation of several non-polar compounds employing a totally organic mobile phase. Once the influence of the evaporative light scattering detector (ELSD) variables on the sensitivity was optimized, the TGs separation was compared conditioning the column with either a conventional HPLC or a MW oven. Contrary to previous applications in which the mobile phase contained water, the improvement in sensitivity using MW was not as significant in comparison with conventional heating but it allowed a shortening in the retention times of several TGs in about 50% respect elution at room temperature. The method was finally applied for the quantification of most common TGs in almond, tiger nut, and argan oil.
Product Description
Supelco
Triglyceride Mix, C2 - C10, ampule of 100 mg (Neat)
Pricing and availability is not currently available.
Triolein, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Tricaprylin, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Trilaurin, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.