MilliporeSigma
  • Lowering circulating apolipoprotein E levels improves aged bone fracture healing.
Lowering circulating apolipoprotein E levels improves aged bone fracture healing.
JCI insight (2019-09-20)
Rong Huang, Xiaohua Zong, Puviindran Nadesan, Janet L Huebner, Virginia B Kraus, James P White, Phillip J White, Gurpreet S Baht
ABSTRACT
Age is a well-established risk factor for impaired bone fracture healing. Here, we identify a role for apolipoprotein E (ApoE) in age-associated impairment of bone fracture healing and osteoblast differentiation, and we investigate the mechanism by which ApoE alters these processes. We identified that, in both humans and mice, circulating ApoE levels increase with age. We assessed bone healing in WT and ApoE-/- mice after performing tibial fracture surgery: bone deposition was higher within fracture calluses from ApoE-/- mice. In vitro recombinant ApoE (rApoE) treatment of differentiating osteoblasts decreased cellular differentiation and matrix mineralization. Moreover, this rApoE treatment decreased osteoblast glycolytic activity while increasing lipid uptake and fatty acid oxidation. Using parabiosis models, we determined that circulating ApoE plays a strong inhibitory role in bone repair. Using an adeno-associated virus-based siRNA system, we decreased circulating ApoE levels in 24-month-old mice and demonstrated that, as a result, fracture calluses from these aged mice displayed enhanced bone deposition and mechanical strength. Our results demonstrate that circulating ApoE as an aging factor inhibits bone fracture healing by altering osteoblast metabolism, thereby identifying ApoE as a new therapeutic target for improving bone repair in the elderly.
Sigma-Aldrich
Sulfobromophthalein disodium salt hydrate, used to study hepatocyte transport functions
Sigma-Aldrich
Fatty Acid Uptake Kit, sufficient for 100 fluorometric tests