MilliporeSigma
  • Development of Immobilized Enzyme Reactors for the characterization of the glycosylation heterogeneity of a protein.
Development of Immobilized Enzyme Reactors for the characterization of the glycosylation heterogeneity of a protein.
Talanta (2019-09-14)
Stan Perchepied, Nicolas Eskenazi, Chiara Giangrande, Julien Camperi, Thierry Fournier, Joëlle Vinh, Nathalie Delaunay, Valérie Pichon
ABSTRACT
The mapping of post-translational modifications (PTMs) of proteins can be addressed by bottom-up proteomics strategy using proteases to achieve the enzymatic digestion of the biomolecule. Glycosylation is one of the most challenging PTM to characterize due to its large structural heterogeneity. In this work, two Immobilized Enzyme Reactors (IMERs) based on trypsin and pepsin protease were used for the first time to fasten and improve the reliability of the specific mapping of the N-glycosylation heterogeneity of glycoproteins. The performance of the supports was evaluated with the digestion of human Chorionic Gonadotropin hormone (hCG), a glycoprotein characterized by four N- and four O-glycosylation sites, prior to the analysis of the digests by nanoliquid chromatography coupled to tandem mass spectrometry (nanoLC-MS/MS). Firstly, the repeatability of the nanoLC-MS/MS was evaluated and a method to control the identification of the identified glycans was developed to validate them regarding the retention time of glycopeptides in reversed phase nanoLC separation. The repeatability of the digestion with trypsin-based IMER was evaluated on the same hCG batch and on three independent batches with common located glycans up to 75%. Then, the performance of the IMER digestions was compared to in-solution digestions to evaluate the qualitative mapping of the glycosylation. It has given rise to 42 out of 45 common glycans between both digestions modes. For the first time, the complementarity of trypsin and pepsin was illustrated for the glycosylation mapping as trypsin led to identifications on 2 out of 4 glycosylation site while pepsin was informative on the 4 glycosylation site. The potential of IMERs for the study of the glycosylation of a protein was illustrated with the comparison of two hCG-based drugs, Ovitrelle® and Pregnyl®.
Sigma-Aldrich
Nα-Benzoyl-L-arginine ethyl ester hydrochloride, trypsin substrate
Sigma-Aldrich
Sodium azide, ≥99%, ultra dry