MilliporeSigma
  • Chronic Treatment With Acetylcholinesterase Inhibitors Attenuates Vascular Dysfunction in Spontaneously Hypertensive Rats.
Chronic Treatment With Acetylcholinesterase Inhibitors Attenuates Vascular Dysfunction in Spontaneously Hypertensive Rats.
American journal of hypertension (2019-03-16)
Renata M Lataro, Marcondes A B Silva, Fabiola L Mestriner, Stefany B A Cau, Rita C A Tostes, Helio C Salgado
ABSTRACT
Acetylcholinesterase inhibition prevents autonomic imbalance, reduces inflammation, and attenuates the development of hypertension. Considering that vascular dysfunction is a crucial feature of arterial hypertension, we investigated the effects of chronic administration of acetylcholinesterase inhibitors-pyridostigmine or donepezil-on vascular reactivity of spontaneously hypertensive rats (SHR). Endothelium-dependent relaxant responses to acetylcholine (ACh) and contractile responses induced by electric field stimulation (EFS) and alpha-adrenergic agonist were studied in mesenteric resistance arteries from SHR and Wistar Kyoto rats. SHR were treated for 16 weeks with vehicle, pyridostigmine (1.5 mg/kg/day) or donepezil (1.4 mg/kg/day). Pyridostigmine and donepezil decreased the vasoconstrictor responses to EFS, which were increased in vehicle-treated SHR. Acetylcholinesterase inhibition increased the modulatory effects of nitric oxide (NO) on SHR vascular reactivity, that is, N(ω)-nitro-(L)-arginine methyl ester (L-NAME) increased EFS-induced contractions and reduced ACh-induced relaxation, with more significant effects in pyridostigmine- and donepezil-treated SHR. The acetylcholinesterase inhibitors also decreased vascular reactive oxygen species levels. This study demonstrates for the first time that long-term administration of acetylcholinesterase inhibitors, pyridostigmine or donepezil, attenuates vascular reactivity dysfunction in SHR by decreasing reactive oxygen species generation and increasing NO bioavailability; possibly via increased endothelial NO synthase activity, and inhibition of NADPH oxidase activity.