MilliporeSigma
  • Adult sox10+ Cardiomyocytes Contribute to Myocardial Regeneration in the Zebrafish.
Adult sox10+ Cardiomyocytes Contribute to Myocardial Regeneration in the Zebrafish.
Cell reports (2019-10-24)
Marcos Sande-Melón, Inês J Marques, María Galardi-Castilla, Xavier Langa, María Pérez-López, Marius-Alexandru Botos, Héctor Sánchez-Iranzo, Gabriela Guzmán-Martínez, David Miguel Ferreira Francisco, Dinko Pavlinic, Vladimir Benes, Rémy Bruggmann, Nadia Mercader
ABSTRACT
During heart regeneration in the zebrafish, fibrotic tissue is replaced by newly formed cardiomyocytes derived from preexisting ones. It is unclear whether the heart is composed of several cardiomyocyte populations bearing different capacity to replace lost myocardium. Here, using sox10 genetic fate mapping, we identify a subset of preexistent cardiomyocytes in the adult zebrafish heart with a distinct gene expression profile that expanded after cryoinjury. Genetic ablation of sox10+ cardiomyocytes impairs cardiac regeneration, revealing that these cells play a role in heart regeneration.
Sigma-Aldrich
Dipeptidyl Peptidase III human, recombinant, expressed in Sf9 cells
Sigma-Aldrich
5-Bromo-2′-deoxyuridine, ≥99% (HPLC)
Supelco
Metronidazole, analytical standard
Sigma-Aldrich
3,3′-Diiodo-L-thyronine (T2) hydrochloride, 98% (CP)
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Sigma-Aldrich
2,3-Butanedione monoxime, ≥98%
Sigma-Aldrich
Anti-Tyrosine Hydroxylase antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution, sufficient for 10 blots