MilliporeSigma
  • Vascular Cells and Tissue Constructs Derived from Human Pluripotent Stem Cells for Toxicological Screening.
Vascular Cells and Tissue Constructs Derived from Human Pluripotent Stem Cells for Toxicological Screening.
Stem cells and development (2019-08-10)
Drew M Titmarsh, Victor Nurcombe, Christine Cheung, Simon M Cool
ABSTRACT
The ability of human stem cells to generate somatic cell lineages makes them ideal candidates for use in toxicological testing and eventually, preclinical drug development. Such resources would support an evolution away from human primary cells or research animal models, which suffer from variability and poor predictability, toward off-the-shelf assays of chemical toxicity and drug efficacy using human cells and tissues. To this end, we generated vascular cell populations (smooth muscle cells and endothelial cells) from human pluripotent stem cells (hPSCs), arranged them into 3D co-cultures within supportive gel matrices, and directed their propensity for self-organization resembling microvasculature. The resulting vascular cell populations and co-cultured constructs were then arrayed in high throughput and used for screening a library of environmental and clinical chemical agents for immunological and toxicological responses. The screen effectively stratified the chemicals into various levels of toxicity, with both cell type-specific and co-culture-dependent responses observed. Thus, hPSC-derived vascular cells and constructs could be progressed further toward use in toxicant and drug screening.
Sigma-Aldrich
BIO, ≥98% (HPLC)
Sigma-Aldrich
Sonepiprazole hydrate, ≥98% (HPLC)
Sigma-Aldrich
Bovine Serum Albumin, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
Supelco
Sildenafil Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
CP-66713, ≥98% (HPLC)