MilliporeSigma
  • Allicin Inhibited Staphylococcus aureus -Induced Mastitis by Reducing Lipid Raft Stability via LxRα in Mice.
Allicin Inhibited Staphylococcus aureus -Induced Mastitis by Reducing Lipid Raft Stability via LxRα in Mice.
Journal of agricultural and food chemistry (2019-09-12)
Yu Chen, Ying Wang, Mei Yang, Meng-Yao Guo
ABSTRACT
Mastitis, inflammation of the mammary gland, occurs in both humans and animals. Staphylococcus aureus is the most common infectious bacterial pathogen associated with mastitis. We investigated the effects of allicin on S. aureus-induced mastitis in mice. Pathological histology revealed that allicin inhibited S. aureus-induced pathological damage and myeloperoxidase activity in mammary tissues. Enzyme-linked immunosorbent assays demonstrated that allicin reduced the production of IL-1β and TNF-α as well as inhibited the NF-κB and mitogen-activated protein kinase pathway by reducing phosphorylation of p65, IκBα, p38, JNK, and ERK. Western blotting revealed that allicin reduced TLR2 and TLR6 expression in mammary tissues and cells but not in HEK293 cells. The lipid raft content was reduced by allicin, which inhibited signaling downstream of TLR2 and TLR6. Liver X receptor α (LXRα) luciferase reporter assays and LXRα interference experiments showed that allicin improved the LXRα activity and adenosine 5'-triphosphate-binding cassette G and A1 (ABCG and ABCA1) expression, thereby reducing the cholesterol level, lipid raft formation, and downstream TLR2 and TLR6 pathway activity. These results demonstrated that allicin exerted anti-inflammatory effects against S. aureus mastitis by improving the LXRα activity and reducing lipid raft formation.
Sigma-Aldrich
Fatty Acid Extraction Kit, Low Standard, sufficient for 40 extractions