MilliporeSigma
  • Enzyme Modification and Oxidative Cross-linking Using Carboxylate-, Phenol- and Catechol-Conjugated 1,8-Naphthalimides.
Enzyme Modification and Oxidative Cross-linking Using Carboxylate-, Phenol- and Catechol-Conjugated 1,8-Naphthalimides.
Photochemistry and photobiology (2019-04-18)
Stacey Sova, Lisa A Kelly
ABSTRACT
The ground- and excited-state interactions of β-alanine, tyrosine and l-dopa substituted 1,8 naphthalimides (NI-Ala, NI-Tyr and NI-Dopa) with lysozyme and mushroom tyrosinase were evaluated to understand the mechanism of oxidative modification. Photooxidative cross-linking of lysozyme was observed for all three conjugates. The yield was significantly reduced for NI-Tyr and NI-Dopa due to intramolecular electron transfer to the excited singlet state of the 1,8-naphthalimide. Incubation of NI-Tyr and NI-Dopa with mushroom tyrosinase resulted in an increased fluorescence from the naphthalimide, suggesting that the phenol and catechol portion of the conjugates are oxidized by the enzyme. This result demonstrates that the compounds bind in the active site of mushroom tyrosinase. The catalytic activity of mushroom tyrosinase to oxidize both tyrosine (monophenolase) and l-dopa (diphenolase) was modified by NI-Tyr and NI-Dopa. Monophenolase activity was inhibited, and the diphenolase activity was enhanced in the presence of these conjugates. Detailed Michaelis-Menten studies show that both Vmax and Km are modified, consistent with a mixed inhibition mechanism. Collectively, the results show that the compounds interact in the enzyme's active site, but also modify the distribution of the enzyme's oxidation states that are responsible for catalysis.
Sigma-Aldrich
1,8-Naphthalic anhydride