MilliporeSigma
  • Azilsartan causes natriuresis due to its sympatholytic action in kidney disease.
Azilsartan causes natriuresis due to its sympatholytic action in kidney disease.
Hypertension research : official journal of the Japanese Society of Hypertension (2019-05-30)
Satoshi Kidoguchi, Naoki Sugano, Koki Takane, Yasuhito Takahashi, Norihiko Morisawa, Miki Yarita, Naomi Hayashi-Ishikawa, Goro Tokudome, Takashi Yokoo
ABSTRACT
The sympathoinhibitory mechanism of azilsartan was investigated in an adenine-induced chronic renal failure model. Azilsartan exerted an antihypertensive effect, though BP elevation induced by adenine was marginal. The creatinine value was significantly lower in the azilsartan group (AZ) than in the vehicle group (VEH); furthermore, proteinuria was suppressed, and sodium excretion was augmented in the AZ group. The low frequency (LF) of systolic BP was suppressed (VEH: 4.07 ± 2.67 mmHg2 vs. AZ: 3.32 ± 1.93 mmHg2 P < 0.001), and the spontaneous baroreflex gain (sBRG) was augmented (VEH: 1.04 ± 0.62ms/mmHg vs. AZ: 1.38 ± 0.69 ms/mmHg, P < 0.001) in AZ. There were no significant differences in ACE1 and ACE2 expression between the groups, which indicated that the action of azilsartan on these components of the intrarenal renin-angiotensin-aldosterone system was comparatively small. Although NHE3, NKCC, and ENaC expression was similar between the groups, NaCl cotransporter (NCC) expression was markedly suppressed by azilsartan (P < 0.05). Thus, in a mild chronic kidney disease (CKD) model with slight BP elevation, the sympatholytic effect of ARB might be expected, and azilsartan might exert its natriuretic effect by NCC suppression achieved by sympathoinhibitory activity.
Sigma-Aldrich
Azilsartan, ≥98% (HPLC)