MilliporeSigma
  • Tissue-specific FAH deficiency alters sleep-wake patterns and results in chronic tyrosinemia in mice.
Tissue-specific FAH deficiency alters sleep-wake patterns and results in chronic tyrosinemia in mice.
Proceedings of the National Academy of Sciences of the United States of America (2019-10-16)
Shuzhang Yang, Sandra M Siepka, Kimberly H Cox, Vivek Kumar, Marleen de Groot, Yogarany Chelliah, Jun Chen, Benjamin Tu, Joseph S Takahashi
ABSTRACT
Fumarylacetoacetate hydrolase (FAH) is the last enzyme in tyrosine catabolism, and mutations in the FAH gene are associated with hereditary tyrosinemia type I (HT1 or TYRSN1) in humans. In a behavioral screen of N-ethyl-N-nitrosourea mutagenized mice we identified a mutant line which we named "swingshift" (swst, MGI:3611216) with a nonsynonymous point mutation (N68S) in Fah that caused age-dependent disruption of sleep-wake patterns. Mice homozygous for the mutation had an earlier onset of activity (several hours before lights off) and a reduction in total activity and body weight when compared with wild-type or heterozygous mice. Despite abnormal behavioral entrainment to light-dark cycles, there were no differences in the period or phase of the central clock in mutant mice, indicating a defect downstream of the suprachiasmatic nucleus. Interestingly, these behavioral phenotypes became milder as the mice grew older and were completely rescued by the administration of NTBC [2-(2-nitro-4-trifluoromethylbenzoyl)-1,3-cyclohexanedione], an inhibitor of 4-hydroxyphenylpyruvate dioxygenase, which is upstream of FAH. Mechanistically, the swst mutation had no effect on the enzymatic activity of FAH, but rather promoted the degradation of the mutant protein. This led to reduced FAH protein levels and enzymatic activity in the liver and kidney (but not the brain or fibroblasts) of homozygous mice. In addition, plasma tyrosine-but not methionine, phenylalanine, or succinylacetone-increased in homozygous mice, suggesting that swst mutants provide a model of mild, chronic HT1.
Sigma-Aldrich
Homogentisic acid, crystalline
Sigma-Aldrich
Nitisinone, ≥95% (HPLC)
Supelco
Nitisinone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Triton X-100, laboratory grade
Millipore
Glutathione−Agarose, lyophilized powder
Sigma-Aldrich
N-Nitroso-N-ethylurea, ISOPAC®