MilliporeSigma
  • A Phase 0 Trial of Ribociclib in Recurrent Glioblastoma Patients Incorporating a Tumor Pharmacodynamic- and Pharmacokinetic-Guided Expansion Cohort.
A Phase 0 Trial of Ribociclib in Recurrent Glioblastoma Patients Incorporating a Tumor Pharmacodynamic- and Pharmacokinetic-Guided Expansion Cohort.
Clinical cancer research : an official journal of the American Association for Cancer Research (2019-07-10)
An-Chi Tien, Jing Li, Xun Bao, Alanna Derogatis, Seongho Kim, Shwetal Mehta, Nader Sanai
ABSTRACT
CDK4/6-dependent cell-cycle regulation is disrupted in most glioblastomas. This study assesses the central nervous system (CNS) pharmacokinetics and tumor pharmacodynamics of ribociclib, a highly selective CDK4/6 inhibitor, in patients with recurrent glioblastoma. Patients with recurrent glioblastoma with intact retinoblastoma protein (RB) expression and CDKN2A deletion or CDK4/6 amplification were treated with ribociclib daily (900 mg) for 5 days before tumor resection. Blood, tumor, and cerebrospinal fluid (CSF) samples were collected, and total and unbound ribociclib concentrations were determined. Pharmacodynamic effects, assessed by RB and FOXM1 phosphorylation, were compared with matched archival tissue. Patients with positive pharmacokinetic and pharmacodynamic effects were enrolled into the expansion cohort for preliminary assessment of progression-free survival (PFS). Twelve patients were enrolled. The mean unbound ribociclib concentrations in CSF, nonenhancing, and enhancing tumor regions were 0.374 μmol/L, 0.560, and 2.152 μmol/kg, respectively, which were more than 5-fold the in vitro IC50 for inhibition of CDK4/6 (0.04 μmol/L). G1-to-S phase suppression was inferred by decreases in phosphorylation of RB (P < 0.01) and cellular proliferation (P < 0.05). Six of 12 patients were enrolled into the pharmacokinetic/pharmacodynamic-guided expansion cohort and demonstrated a median PFS of 9.7 weeks. Examination of recurrent tumors following monotherapy indicated upregulation of the PI3K/mTOR pathway. Ribociclib exhibited good CNS penetration, and target modulation was indicated by inhibition of RB phosphorylation and tumor proliferation. Ribociclib monotherapy showed limited clinical efficacy in patients with recurrent glioblastoma. Combination therapy with CDK4/6 and PI3K/mTOR inhibitors may be explored for treating recurrent glioblastoma.
Sigma-Aldrich
Anti-FOXM1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution