MilliporeSigma
  • A combined experimental and computational study on peptide nucleic acid (PNA) analogues of tumor suppressive miRNA-34a.
A combined experimental and computational study on peptide nucleic acid (PNA) analogues of tumor suppressive miRNA-34a.
Bioorganic chemistry (2019-08-17)
Valerio Piacenti, Emma Langella, Ida Autiero, John C Nolan, Olga Piskareva, Mauro F A Adamo, Michele Saviano, Maria Moccia
ABSTRACT
MicroRNAs are a ubiquitous class of non-coding RNAs able to regulate gene expression in diverse biological processes. Widespread miRNAs deregulation was reported in numerous diseases including cancer, with several miRNAs playing oncogenic and/or tumor suppressive role by targeting multiple mRNAs simultaneously. Based on these findings, miRNAs have emerged as promising therapeutic tools for cancer treatment. Herein, for the first time, peptide nucleic acids (PNAs) were studied to develop a new class of molecules able to target 3'UTR on MYCN mRNA without a fully complementary base pairing sequence (as miRNAs). For our proof of concept study we have selected as a model the miRNA-34a, which acts as a tumor suppressor in a number of cancers including neuroblastoma. In particular, miRNA-34a is a direct regulator of MYCN oncogene, whose overexpression is a prominent biomarker for the highly aggressive neuroblastoma phenotype. The design and synthesis of three PNA-based oligomers of different length was described, and their interaction with two binding sites on the target MYCN mRNA was investigated by molecular dynamics simulation, and spectroscopic techniques (CD, UV). Intake assay and confocal microscopy of PNA sequences were also carried out in vitro on neuroblastoma Kelly cells. Despite the presence of multiple mismatches, the PNA/RNA hetero duplexes retain very interesting features in terms of stability, affinity as well as of cellular uptake.
Sigma-Aldrich
Fmoc-Lys(Boc)-OH, ≥98.0% (HPLC)
Sigma-Aldrich
Fmoc-Cys(Trt)-OH, ≥95.0% (sum of enantiomers, HPLC)